Learn More
Environmental Express™ Snip & Pour™ Standards
Catalog No. 09735152 Shop All Environmental Express Products
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
09-735-152 10 mL
1 options
Catalog No. 09-735-152 Supplier Environmental Express™ Supplier No. G3025
Add to Cart
Add to Cart
Requires
Manifold, SPE extraction disk and Na2SO4 drying cartridge must be purchased separately.
Premeasured controls for oil and grease analysis
Simplifies U.S. EPA Method 1664
  • Meets requirements for EPA Method 1664
  • Each lot is quality checked gravitmetrically and by GC
    Specifications
    For Use With (Application) Solid Phase Extraction
    Product Line ENVIRONMENTAL EXPRESS
    Type HEM-Filled PTFE Tubes
    Volume (Metric) 10 mL
    Material PTFE