Learn More
Environmental Express™ MaxFil/ZHE+ Pressure Filtration Replacement Parts
Catalog No. 0973529 Shop All Environmental Express Products
Change view
Click to view available options
Material:
Viton™-A
Catalog No. Material
09-735-29 Viton™-A
1 options
Catalog No. 09-735-29 Supplier Environmental Express™ Supplier No. ZVR9903
Add to Cart
Add to Cart
Extend the life of your MaxFil/ZHE+ Pressure Filtration Systems with these replacement parts
  • MaxFil Pressure Filtration unit parts are available
  • Zero Headspace Extractor ZHE+ unit parts are available
Specifications
Type O-ring Set
Material Viton™-A
For Use With (Equipment) ZHE Plug Valve