Learn More
Environmental Slide Set 3: Life in the Soil
SCI_ED
Catalog No. S64944 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Type:
Our Environ Slide Set
Catalog No. Type
S64944 Our Environ Slide Set
1 options
Catalog No. S64944 Supplier Johannes Lieder GmbH & Co. KG Supplier No. 4570
Add to Cart
Add to Cart
Includes
  • Acidophils soil bacteria, solution of heavy metals
  • Nitrite bacteria, forming harmful nitrogenous substances
  • Root of Beech with ectotrophic mycorrhiza
  • Root of Birch with partly endotropic mycorrhiza
  • Netted venation, portion of rotted deciduous leaf
  • Stem of green manure plant
  • Soil bacteria (Bacillus megaterium) smear
  • Hyphae of root fungi
  • Lichen, indicator of clean air
  • Mushroom mycelium
  • Root of Willow plant — used to protect against erosion
  • Earthworm — causes soil improvement
  • Springtails (Collembola)
  • Mite from forest soil
  • Constituents of humus soil
  • Constituents of peat soil
Take a microscopic look at life in the soil.
Set of 17 slides details microscopic activity in soil.
Specifications
Type Our Environ Slide Set