Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Environstar MP Coveralls

Catalog No. 19161128D Shop All Tians International Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Neck Style:
Collared
Hooded
Waist Style:
Elastic
Non-elastic
Ankle Style:
Elastic
Elastic, Attached Boots
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Neck Style Waist Style Ankle Style
19-161-128D X-Large Collared Elastic Elastic
19-130-200E 3X-Large Collared Non-elastic -
19-130-200C X-Large Collared Non-elastic -
19-130-200F 4X-Large Collared Non-elastic -
19-130-200D 2X-Large Collared Non-elastic -
19-130-201C X-Large Hooded Elastic Elastic
19-166-140 3X-Large Hooded Elastic Elastic, Attached Boots
19-161-128E 2X-Large Collared Elastic Elastic
19-166-400 4X-Large Hooded Elastic Elastic, Attached Boots
19-161-128C Large Collared Elastic Elastic
19-130-201B Large Hooded Elastic Elastic
19-161-128G 4X-Large Collared Elastic Elastic
19-130-201G 5X-Large Hooded Elastic Elastic
19-130-201D 2X-Large Hooded Elastic Elastic
19-161-128H 5X-Large Collared Elastic Elastic
19-130-200B Large Collared Non-elastic -
19-130-201E 3X-Large Hooded Elastic Elastic
19-130-201F 4X-Large Hooded Elastic Elastic
19-161-128B Medium Collared Elastic Elastic
19-130-201A Medium Hooded Elastic Elastic
19-130-200A Medium Collared Non-elastic -
19-161-128F 3X-Large Collared Elastic Elastic
19-161-128A Small Collared Elastic Elastic
19-130-200G 5X-Large Collared Non-elastic -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Catalog No. 19-161-128D Supplier Tians Supplier No. 216853XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protective apparel manufactured in climate-controlled facility

Specifications

Product Type Cleanroom Coveralls
Size X-Large
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged
Coating Microporous Polypropylene
Collar Type Standard fold
Certifications/Compliance ISO Class 6, ANSI ISEA 101-1996
For Use With (Application) Protects against petroleum Oils, pesticides, Bleach and Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.