Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien EnviroStar™ Noninfectious Waste Sharps Disposal Containers

Catalog No. 14830107
Change view
Click to view available options
Capacity (English):
2 gal.
8 gal.
18 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
14-830-107 2 gal.
14-830-108 8 gal.
14-830-110 18 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-830-107 Supplier Covidien Supplier No. 8790
This item is not returnable. View return policy

Manufactured with recyled materials, these containers are designed to reduce injuries and disposal costs for non-contaminated sharps

  • Designed to contain sharps that can cause a percutaneous tear but are not infected with bloodborne pathogens, such as light bulbs, broken slides or Petri dishes, and glass lab bottles
  • Environmental green container color helps to differentiate from other disposal containers and promotes proper waste segregation
  • Made with environmentally friendly recycled material, enhancing Reduce-Recycle-Reuse programs

Specifications

Capacity (Metric) 7.6 L
Capacity (English) 2 gal.
Color Green
Model EnviroStar
Autoclavable Autoclavable
For Use With (Application) For sharps not infected with blood-borne pathogens
Material Recycled Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.