Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534959 Supplier Invitrogen™ Supplier No. PA534959
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (86%), Xenopus laevis (93%), Pig (100%), Chicken (96%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ENY2 is a component of the transcription regulatory histone acetylation (HAT) complex SAGA, a multiprotein complex that activates transcription by remodeling chromatin and mediating histone acetylation and deubiquitination. Within the SAGA complex, participates in a subcomplex that specifically deubiquitinates both histones H2A and H2B. The SAGA complex is recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation. May also participate in mRNA export and accurate chromatin positioning in the nucleus by tethering genes to the nuclear periphery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENY2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ENY2
Gene Accession No. Q9NPA8
Gene Alias 1810057B09Rik; 6720481I12; DC6; e(y)2; e(y)2 homolog; e(y)2 protein; enhancer of yellow 2 homolog; enhancer of yellow 2 homolog (Drosophila); enhancer of yellow 2 transcription factor homolog; enhancer of yellow eye homolog; ENY 2; ENY2; ENY-2; ENY2, transcription and export complex 2 subunit; Ey2; Sus1; transcription and mRNA export factor ENY2
Gene Symbols ENY2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ENY2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.