Learn More
Selleck Chemical LLC Enzastaurin (LY317615) 10mg 170364-57-5
SDP
Supplier:  Selleck Chemical LLC S105510MG
Encompass_Preferred
Enzastaurin (LY317615) is a potent PKC? selective inhibitor with IC50 of 6 nM in cell-free assays, 6- to 20-fold selectivity against PKC?, PKC? and PKC?. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-834-6