Learn More
Bioassay Systems EnzyChrom™ Acetaldehyde Assay Kit. For the quantitative determination of acetaldehyde in biological and food/beverage samples
SDP
Catalog No. NC1317896 Shop All Bioassay Systems Products
Encompass_Preferred
For the quantitative determination of acetaldehyde in biological and food/beverage samples. Method: OD565nm; Samples: Biological, food and beverage samples; Size: 100 tests; Detection Limit: 2 µM. Key Features: Fast and sensitive. Linear detection range (20µL sample): 2µM to 2 mM acetaldehyde in 96-well plate assay. High-throughput. The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Room temperature assay. No 37°C heater is needed. Convenient. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day.
Catalog No. NC1317896 Supplier Bioassay Systems Supplier No. EACT100
For the quantitative determination of acetaldehyde in biological and food/beverage samples. Method: OD565nm; Samples: Biological, food and beverage samples; Size: 100 tests; Detection Limit: 2 µM. Key Features: Fast and sensitive. Linear detection range (20µL sample): 2µM to 2 mM acetaldehyde in 96-well plate assay. High-throughput. The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Room temperature assay. No 37°C heater is needed. Convenient. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day.