Learn More
Bioassay Systems EnzyChrom™ a-Amylase Assay Kit. For quantitative determination of a-amylase activity.
SDP
Catalog No. 50489251 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of α-amylase enzyme activity. Key Features: Sensitive and accurate. Linear detection range 0.3 to 50 U/L α-amylase in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, incubation for 15 min, followed by the detection reagent and a 20-min incubation and reading the optical density at 585 nm. Method: OD585nm. Samples: Blood, saliva, urine, agriculture etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.3 U/L.
Catalog No. 50-489-251 Supplier Bioassay Systems Supplier No. ECAM100
Add to Cart
Add to Cart
For quantitative determination of α-amylase enzyme activity. Key Features: Sensitive and accurate. Linear detection range 0.3 to 50 U/L α-amylase in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, incubation for 15 min, followed by the detection reagent and a 20-min incubation and reading the optical density at 585 nm. Method: OD585nm. Samples: Blood, saliva, urine, agriculture etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.3 U/L.