Learn More
Bioassay Systems EnzyChrom™ Choline Assay Kit. For quantitative determination of choline and evaluation of drug effects on choline metabolism.
SDP
Catalog No. 50489255 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of choline and evaluation of drug effects on choline metabolism. Key Features: Use 20 µL samples. Linear detection range: colorimetric assay 1 to 100 µM, fluorimetric assay 0.2 to 10 µM choline. Method: OD570nm, or FL530/585nm. Samples: Serum, plasma, urine, saliva, milk, tissue, cells etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: OD, FL: 1, 0.2 µM.
Catalog No. 50-489-255 Supplier Bioassay Systems Supplier No. ECHO100
Add to Cart
Add to Cart
For quantitative determination of choline and evaluation of drug effects on choline metabolism. Key Features: Use 20 µL samples. Linear detection range: colorimetric assay 1 to 100 µM, fluorimetric assay 0.2 to 10 µM choline. Method: OD570nm, or FL530/585nm. Samples: Serum, plasma, urine, saliva, milk, tissue, cells etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: OD, FL: 1, 0.2 µM.