Learn More
Bioassay Systems EnzyChrom™ Creatine Assay Kit. For quantitative determination of creatine and evaluation of drug effects on creatine metabolism.
SDP
Catalog No. 50489261 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of creatine and evaluation of drug effects on creatine metabolism. Key Features: High sensitivity and wide linear range. Use 10 µL sample. Linear detection range 4 to 1000 µM (colorimetric) or 0.5 to 50 µM (fluorimetric). Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of creatine within 30 minutes. Method: OD570nm, or FL530/590nm. Samples: Serum, plasma, urine, saliva etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 4 µM.
Catalog No. 50-489-261 Supplier Bioassay Systems Supplier No. ECRT100
Add to Cart
Add to Cart
For quantitative determination of creatine and evaluation of drug effects on creatine metabolism. Key Features: High sensitivity and wide linear range. Use 10 µL sample. Linear detection range 4 to 1000 µM (colorimetric) or 0.5 to 50 µM (fluorimetric). Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of creatine within 30 minutes. Method: OD570nm, or FL530/590nm. Samples: Serum, plasma, urine, saliva etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 4 µM.