Learn More
Bioassay Systems EnzyChrom™ Glycogen Assay Kit. For quantitative determination of glycogen and evaluation of drug effects on glycogen metabolism.
SDP
Catalog No. 50489237 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of glycogen and evaluation of drug effects on glycogen metabolism. Key Features: Use as little as 10 µL samples. Linear detection range: 2 to 200 µg/mL glycogen for colorimetric assays and 0.2 to 20 µg/mL for fluorimetric assays. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: OD, FL: 2, 0.2 µg/mL.
Catalog No. 50-489-237 Supplier Bioassay Systems Supplier No. E2GN100
For quantitative determination of glycogen and evaluation of drug effects on glycogen metabolism. Key Features: Use as little as 10 µL samples. Linear detection range: 2 to 200 µg/mL glycogen for colorimetric assays and 0.2 to 20 µg/mL for fluorimetric assays. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: OD, FL: 2, 0.2 µg/mL.