Learn More
Bioassay Systems EnzyChrom™ L-Arginine Assay Kit, For quantitative determination of L-arginine in food, beverage, biological samples (e.g. serum, cell lysate, etc).
SDP
Catalog No. 5000144367 Shop All Bioassay Systems Products
Encompass_Preferred
L-ARGININE is an amino acid essential for protein metabolism and primarily found in the active sites of various proteins. During nitrogen metabolism, arginase breaks down arginine into urea and ornithine. L-Arginine is also present in many protein-rich foods and beverages. In particular, its detection in beverages, such as juices and wine, is crucial for quality control. BioAssay Systems’ L-Arginine assay measures the urea generated from arginase hydrolysis of arginine with our proprietary reagents. The increase in absorbance at 450 nm is proportional to the arginine concentration.
Catalog No. 50-001-44367 Supplier Bioassay Systems Supplier No. EAGN100
L-ARGININE is an amino acid essential for protein metabolism and primarily found in the active sites of various proteins. During nitrogen metabolism, arginase breaks down arginine into urea and ornithine. L-Arginine is also present in many protein-rich foods and beverages. In particular, its detection in beverages, such as juices and wine, is crucial for quality control. BioAssay Systems’ L-Arginine assay measures the urea generated from arginase hydrolysis of arginine with our proprietary reagents. The increase in absorbance at 450 nm is proportional to the arginine concentration.