Learn More
Bioassay Systems EnzyChrom™ Lactose Assay Kit. For quantitative determination of lactose and evaluation of drug effects on lactose metabolism.
SDP
Catalog No. 50489278 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of lactose and evaluation of drug effects on lactose metabolism. Key Features: Sensitive. Use as little as 20 µL samples. Linear detection range in 96-well plate: 17 to 2000 µM lactose for colorimetric assays and 6 to 100 µM for fluorimetric assays. Convenient and high-throughput. Robust and homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological (milk), food and agriculture etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 6 µM.
Catalog No. 50-489-278 Supplier Bioassay Systems Supplier No. ELAC100
For quantitative determination of lactose and evaluation of drug effects on lactose metabolism. Key Features: Sensitive. Use as little as 20 µL samples. Linear detection range in 96-well plate: 17 to 2000 µM lactose for colorimetric assays and 6 to 100 µM for fluorimetric assays. Convenient and high-throughput. Robust and homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological (milk), food and agriculture etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 6 µM.