Learn More
Bioassay Systems EnzyChrom™ Sialic Acid Assay Kit. For quantitative determination of free and total sialic acid (NANA) and evaluation of drug effects on its metabolism.
SDP
Catalog No. 50489291 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of free and total sialic acid (NANA) and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Use as little as 10 µL samples. Linear detection range in 96-well plate: 0.02 to 1 mM sialic acid for colorimetric assays and 2 to 100 µM for fluorimetric assays. Simple and convenient. Can detect free sialic acid by addition of a single working reagent and incubation for 60 min at room temperature or total sialic acid by pre-treating samples with a 60 min hydrolysis step. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 2 hrs. Kit size: 100 tests. Detection limit: OD, FL: 20, 2 µM.
Catalog No. 50-489-291 Supplier Bioassay Systems Supplier No. ESLA100
For quantitative determination of free and total sialic acid (NANA) and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Use as little as 10 µL samples. Linear detection range in 96-well plate: 0.02 to 1 mM sialic acid for colorimetric assays and 2 to 100 µM for fluorimetric assays. Simple and convenient. Can detect free sialic acid by addition of a single working reagent and incubation for 60 min at room temperature or total sialic acid by pre-treating samples with a 60 min hydrolysis step. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 2 hrs. Kit size: 100 tests. Detection limit: OD, FL: 20, 2 µM.