Learn More
Bioassay Systems EnzyChrom™ Starch Assay Kit. For quantitative determination of starch and evaluation of drug effects on starch metabolism.
SDP
Catalog No. 50489240 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of starch and evaluation of drug effects on starch metabolism. Key Features: Use as little as 10 µL samples. Linear detection range: 2 to 200 µg/mL starch for colorimetric assays and 0.2 to 20 µg/mL for fluorimetric assays. Method: OD570nm, or FL530/585nm. Samples: Biological, agriculture, food etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 2 µg/mL.
Catalog No. 50-489-240 Supplier Bioassay Systems Supplier No. E2ST100
For quantitative determination of starch and evaluation of drug effects on starch metabolism. Key Features: Use as little as 10 µL samples. Linear detection range: 2 to 200 µg/mL starch for colorimetric assays and 0.2 to 20 µg/mL for fluorimetric assays. Method: OD570nm, or FL530/585nm. Samples: Biological, agriculture, food etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 2 µg/mL.