Learn More
Bioassay Systems EnzyChrom™ Triglyceride Assay Kit. For quantitative determination of triglyceride and evaluation of drug effects on triglyceride metabolism.
SDP
Catalog No. 50489294 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of triglyceride and evaluation of drug effects on triglyceride metabolism. Key Features: Sensitive and accurate. Use as little as 10 µL samples. Linear detection range 0.01 to 1.0 mmol/L (0.88 to 88.5 mg/dL) triglyceride. Simple and convenient. The procedure involves addition of a single working reagent and incubation for 30 min at room temperature, compatible for HTS assays. Improved reagent stability. The optimized formulation has greatly enhanced the reagent and signal stability. Method: OD570nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 30 min. Kit size: 200 tests. Detection limit: 0.01 mmol/L (0.88 mg/dL).
Catalog No. 50-489-294 Supplier Bioassay Systems Supplier No. ETGA200
For quantitative determination of triglyceride and evaluation of drug effects on triglyceride metabolism. Key Features: Sensitive and accurate. Use as little as 10 µL samples. Linear detection range 0.01 to 1.0 mmol/L (0.88 to 88.5 mg/dL) triglyceride. Simple and convenient. The procedure involves addition of a single working reagent and incubation for 30 min at room temperature, compatible for HTS assays. Improved reagent stability. The optimized formulation has greatly enhanced the reagent and signal stability. Method: OD570nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 30 min. Kit size: 200 tests. Detection limit: 0.01 mmol/L (0.88 mg/dL).