Learn More
Bioassay Systems EnzyFluo™ L-Lactate Assay Kit. For quantitative determination of L-lactate (L-lactic acid) and evaluation of drug effects on its metabolism.
SDP
Catalog No. 50489266 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of L-lactate (L-lactic acid) and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Detection limit of 1 µM and linearity up to 50 µM L-lactate in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the fluorescence after 60 min. Room temperature assay. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: FL530/585 nm. Samples: Serum, plasma, cell culture media etc. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: 1 µM.
Catalog No. 50-489-266 Supplier Bioassay Systems Supplier No. EFLLC100
For quantitative determination of L-lactate (L-lactic acid) and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Detection limit of 1 µM and linearity up to 50 µM L-lactate in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the fluorescence after 60 min. Room temperature assay. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: FL530/585 nm. Samples: Serum, plasma, cell culture media etc. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: 1 µM.