Learn More
Bioassay Systems EnzyLight™ ADP Assay Kit. For rapid, quantitative, bioluminescent determination of ADP concentration and evaluation of drug effects on ADP metabolism.
SDP
Catalog No. NC1255584 Shop All Bioassay Systems Products
Encompass_Preferred
For rapid, quantitative, bioluminescent determination of ADP concentration and evaluation of drug effects on ADP metabolism. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.02 µM ADP can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: Z factors of 0.5 and above are routinely observed in 96-well and 384-well plates. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: Luminescence; Samples: Cells etc; Size: 100 tests.
Catalog No. NC1255584 Supplier Bioassay Systems Supplier No. EADP100
For rapid, quantitative, bioluminescent determination of ADP concentration and evaluation of drug effects on ADP metabolism. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.02 µM ADP can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: Z factors of 0.5 and above are routinely observed in 96-well and 384-well plates. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: Luminescence; Samples: Cells etc; Size: 100 tests.