Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corepoint Scientific™ Enzyme Laboratory Freezer
SDP

Catalog No. 22650677 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
230V 50Hz
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-650-677 230V 50Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 22-650-677 Supplier Corepoint Scientific™ Supplier No. NSZF241WMW/5M

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Requires

Cord and plug for 230V 50Hz model.

Energy efficiency in a 24 cu. ft. freezer with heavy-duty refrigeration system

  • Digital LED display microprocessor temperature controller
  • Remote alarm contacts, and warm and cold audible and visual alarms
  • Adjustable control range from -10°C to -25°C; factory preset at -20°C
  • Continuous product temperature display
  • Product sensor with glycerin bottle
  • Power supply switch for manual defrost
  • Air-cooled condensing unit
  • Centered key-lockable doors (two keys)
  • 1 in. dia. sensor access port

Construction

  • Top-mounted refrigeration system
  • Full-length pull door handle
  • Heavy-duty solid door with spring-loaded hinges and magnetic door gasket
  • Perimeter anti-condensate door frame heater wires
  • Interior cabinet liner and interior door panel are aluminum
  • Exterior cabinet front, sides and back painted white; top and bottom are galvanized steel
  • Cabinet and solid door are foamed in-place with CFC-free high-density polyurethane foam insulation
  • Four casters (two locking)
  • Nine epoxy-coated wire shelves

Order Info

Optional features, including extra shelves, drawers, sensor access port, legs, stainless-steel interior/exterior, reverse door hinge, storage bins, temperature chart recorder, and 4-20ma output, available. Contact your Fisher Scientific™ representative for more information.

Warranty and Services

18 months parts and labor, 5 years compressor (US and Canada); 18 months parts (International)

Specifications

Capacity (English) 24 cu. ft.
Capacity (Metric) 680 L
Temperature Range -20°C
Temperature Sensor Microprocessor
No. of Doors 1
Shelves 9
Certifications/Compliance CE, CEC, cUL, UL
Style Upright
Color White
Defrost Manual
Dimensions (D x W x H) Interior 30 x 23.5 x 59 in. (76 x 59 x 149 cm)
Dimensions (L x W x H) Exterior 34.87 x 28.5 x 79.6 in. (88 x 72 x 202 cm)
Depth (English) Interior 30 in.
Depth (Metric) Interior 76 cm
Height (English) Exterior 79.6 in.
Height (English) Interior 59 in.
Height (Metric) Exterior 202 cm
Height (Metric) Interior 149 cm
Length (English) Exterior 34.87 in.
Length (Metric) Exterior 88 cm
Width (English) Exterior 28.5 in.
Width (English) Interior 23.5 in.
Width (Metric) Exterior 72 cm
Width (Metric) Interior 59 cm
Amperage 15 A
Voltage 230 V
Electrical Requirements 230 V 50 Hz
Frequency 50 Hz
Refrigerant R-404A
Weight (English) 419 lb.
Weight (Metric) 190.05 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.