Learn More
Abcam Enzyme Mix, Lyophilized, 1 ea
Description
Enzyme Mix (lyophilized) supplied for assay use.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Lyophilized |
| Kit Contents | Enzyme Mix: Lyophilized |
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Cholesterol Esterase |
| Quantity | 1 Unit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.