Learn More
Creative Biomart Thyroglobulin; hTG
Supplier:  Creative Biomart THP0036
Encompass_Preferred
The product (also known as Tg) is an enzyme thyroid hormone used by the thyroid gland to produce the thyroid hormones thyroxine (T4) and triiodothyronine (T3). The active form of thyroxine, triiodothyronine, is produced both within the thyroid gland and periphery by 5'-deiodinase.
Catalog No. 50-208-4087
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.