Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Eos Monoclonal Antibody (ESB7C2), PE, eBioscience™, Invitrogen™

Catalog No. 12575880
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-575-880 25 μg
50-112-2332 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-575-880 Supplier Invitrogen™ Supplier No. 12575880
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This ESB7C2 monoclonal antibody reacts with mouse Eos (IKZF4, ZNFN1A4), a zinc finger transcription factor and member of the Ikaros family. Among the Ikaros family of proteins, Eos has the highest homology with Helios. Eos can homodimerize with itself and form heterodimers with Ikaros. Eos was first identified in the nervous system and was cloned from astrocytes. Later, Eos was found to be involved in the regulation of osteoclast differentiation. Eos has also been reported to be involved in negatively regulating gene expression in Foxp3-expressing regulatory T cells in mice. Applications Reported: This ESB7C2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This ESB7C2 antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the using the Foxp3/Transcription Factor Buffer Set (cat. 00-5523) and protocol. Slight modifications in the protocol using this antibody have been demonstrated to improve the staining resolution. We recommend the staining time of minimum 60 minutes and additional washes after the fixation (3 times) and staining steps (twice). This antibody can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Serine protease that has amidolytic activity, cleaving its substrates before Arg residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eos
Applications Flow Cytometry
Classification Monoclonal
Clone ESB7C2
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Prss33
Gene Accession No. Q80WM7
Gene Alias Eos; mT6; protease, serine 33; protease, serine, 33; Prss33; Serine protease 33; serine protease EOS; tryptase 6; Tryptase-6
Gene Symbols Prss33
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353130
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.