Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y, 0.2% AQ, Vion Biosciences
SCI_ED

Catalog No. S02512 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02512 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02512 Supplier Aldon Corporation Supplier No. VNEE0060500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With Eosin Y 0.2% aqueous, stain cytoplasm, red blood cells, collagen, and muscle fibers for histological examination or use as a counterstain with hematoxylin.

  • With Eosin Y 0.2% aqueous, stain cytoplasm, red blood cells, collagen, and muscle fibers for histological examination or use as a counterstain with hematoxylin.

Chemical Identifiers

CAS 17372-87-1 , 7732-18-5
Molecular Formula C20H6Br4Na2O5
Molecular Weight (g/mol) 691.86
MDL Number MFCD00005040
InChI Key SEACYXSIPDVVMV-UHFFFAOYSA-L
Synonym Eosin Y, Water Solution
PubChem CID Data not available.
IUPAC Name disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12

Specifications

Melting Point 0°C
Boiling Point 100°C
CAS Min % 0.20%, 99.80%
CAS Max % 99.80%, 0.20%
Concentration 0.20%
Dye Content 0.20%
Assay Percent Range 0.20%
Extinction Coefficient 112000 M-1cm-1 (at 525 nm)
UN Number Not regulated.
Solubility Information Complete in water.
Identification 0.2 % Eosin Y Aqueous Solution
Infrared Spectrum Yes
Lambda Max 525 nm
Percent Purity 0.20%
Grade Data not available.
Concentration or Composition (by Analyte or Components) 0.2% in Water
Molar Absorption Coefficient 112000 M-1cm-1 (at 525 nm)
Odor No odor.
Packaging HDPE Bottle
Specific Gravity 1.0 g/mL in water)
Vapor Pressure 14 mm Hg in water)
Certificate of Analysis Yes
Decomposition Information Data not available.
Density 1.0 g/mL in water)
pH Color Changes Data not available.
Solvent or Matrix Water
Color Orange
pH Data not available.
Quantity 500 mL
Flash Point Data not available.
Physical Form Data not available.
Chemical Name or Material Eosin Y
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category None.
ShelfLife 3 years
DOTInformation Not regulated.
EINECSNumber 241-409-6
RTECSNumber LM5800000
Recommended Storage Store in a cool, well-ventilated area away from incompatible substances. Protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.