Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y Disodium Salt, MP Biomedicals

Catalog No. ICN15247125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15247125 25 g
ICN15247180 100 g
ICN15247183 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15247125 Supplier MP Biomedicals, Inc Supplier No. 0215247125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym 2',4',5',7'-Tetrabromofluorescein disodium salt, 2', 4', 5', Acid Red 87
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Formula Weight 691.86
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Eosin Y Disodium Salt
Recommended Storage Store at Room Temperature (15 to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.