Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Eosin-Y Phloxine-B, iCON™ Staining System
SDP

Catalog No. 21377137
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
4 x 16 oz.
4 x 1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-137 16 oz.
21-377-138 1 gal.
21-377-139 4 x 1 gal.
21-377-140 4 x 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-377-137 Supplier Azer Scientific Supplier No. ES3653316
Add to Cart
Add to Cart
This item is not returnable. View return policy

Producing crisp and well-defined detail is our proprietary family of pH-balanced

  • Producing crisp and well-defined detail is our proprietary family of pH-balanced iCON™ Intensified Cytoplasmic Stains
  • This family of cytoplasmic stains consists of two distinct products designed to produce consistent quality results
  • Designed to produce consistent quality results
  • May be used on all automatic stainers as well as in manual staining
  • Produces a wide range of color intensities and staining brilliance

Specifications

For Use With (Application) Automatic Stainers
Quantity 16 oz.
Reactivity 1 gal.
Reagent Type 4 x 1 gal.
Sample Type 4 x 16 oz.
Product Type Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.