Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y Stain, 1% (w/v) Alcoholic Stock Solution, Ricca Chemical
SDP

Catalog No. 28501 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2850-1 4 L Natural Poly Bottle
2850-16 500 mL Natural Poly Bottle
2850-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2850-1 Supplier Ricca Chemical Company Supplier No. 28501

This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative.

Specifications

Chemical Name or Material Eosin Y Stain
Name Note Stock Solution
CAS 64-17-5, 17372-87-1, 67-56-1
CAS Min % 97.0, 1.21
CAS Max % 100.0, 1.26
Concentration 1% (w/v)
Percent Purity 1% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Alcoholic Solution
Color Green
Quantity 4 L
Physical Form Murky Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.