Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y Stain, 1% (w/v) Alcoholic Stock Solution, Ricca Chemical
SDP

Catalog No. 285016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2850-16 500 mL Natural Poly Bottle
2850-32 1 L Natural Poly Bottle
2850-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2850-16 Supplier Ricca Chemical Company Supplier No. 285016
Add to Cart
Add to Cart

Specifications

Chemical Name or Material EOSIN Y
CAS 64-17-5, 17372-87-1, 67-56-1
CAS Min % 97.0, 1.21
CAS Max % 100.0, 1.26
Molecular Formula C20H12Br4Na2O8
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
Molecular Weight (g/mol) Mixture
PubChem CID 91886399
Percent Purity 1% W/V
Grade Indicator
Packaging Natural Poly Bottle
Color Green
Quantity 500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.