Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y Yellow Certified Macron Fine Chemicals™

Catalog No. MK0460100 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-0460-100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-0460-100 Supplier Avantor Supplier No. 046057

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym 2,4,5,7-tetrabromogluorescein
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Formula Weight 691.86
Packaging Glass Bottle
Quantity 100 g
Chemical Name or Material Eosin Y

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.