Learn More
Invitrogen™ Eotaxin 2 Polyclonal Antibody, PeproTech®, Invitrogen™
Description
AA Sequence of recombinant protein: VTIPSSCCTS FISKKIPENR VVSYQLANGS ICPKAGVIFI TKKGHKICTD PKLLWVQRHI QKLDAKKNQP SKGAKAVRTK FAVQRRRGNS TEV. Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine Eotaxin-2 (CCL24). Anti-Murine Eotaxin-2 (CCL24)-specific antibody was purified by affinity chromatography employing an immobilized Murine Eotaxin-2 (CCL24) matrix. Sandwich ELISA: To detect Murine Eotaxin-2 (CCL24) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine Eotaxin-2 (CCL24) (500-P175GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine Eotaxin-2 (CCL24). Western Blot: To detect Murine Eotaxin-2 (CCL24) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine Eotaxin-2 (CCL24) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Specifications
Specifications
| Antigen | Eotaxin 2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CCL24 |
| Gene Accession No. | Q9JKC0 |
| Gene Alias | CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 |
| Gene Symbols | CCL24 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.