Learn More
EP Reference Solution BY3, Brownish-Yellow Reference Solution, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Brownish-Yellow Standard Solution |
| Packaging | Amber Glass Bottle |
| CAS | 7647-01-0, 7732-18-5, 10025-77-1, 7758-99-8, 7791-13-1 |
| Physical Form | Liquid |
| Color | Brown or Green/Yellow |
| pH | Acidic |
| Melting Point | 0.0°C |
| Boiling Point | 100°C |
| For Use With (Equipment) | Water Analyzer |
| Solubility | Miscible with water |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.