Learn More
Synthware™ EP™ O-Rings
GREENER_CHOICESDP
Catalog No. 31502359 Shop All Kemtech Products
Change view
Click to view available options
For Use With (Equipment):
#15 GL Thread
#25 GL Thread
#7 GL Thread
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
31-502-359 #15 GL Thread
31-502-361 #7 GL Thread
31-502-360 #25 GL Thread
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-359 Supplier Synthware™ Supplier No. R111EP
Add to Cart
Add to Cart
Long lasting O-rings for solvent and chemical compatibility.
  • O-rings for solvent and chemical compatibility
Specifications
For Use With (Equipment) #15 GL Thread
Product Type O-Ring
Material Ethylene Propylene
Diameter (Metric) Outside 16 mm
Diameter (English) Outside 0.63 in.
Size 111