Learn More
Cayman Chemical EP4 Receptrrat Reporter A 1ea
SDP
Supplier:  Cayman Chemical 6003501 EA
Encompass_Preferred
An Mps1 inhibitor (IC50 0.34 nM) inhibits nocodazole-induced metaphase arrest in HeLa cells (IC50 56 nM) inhibits proliferation in a panel of 86 cancer cell lines (median IC50 6.7 nM) reduces tumor area in a cisplatin-resistant A2780/CP70 ovarian cancer mouse xenograft model at 2.5 mg/kg
Catalog No. 50-289-6848
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.