Learn More
Aviva Systems Biology Corp EP300 Antibody (OAAF01891) | 100 ug
SDP
Supplier:  Aviva Systems Biology Corp OAAF01891
Encompass_Preferred
The antiserum was produced against synthesized peptide derived between 1-50 amino acids from the N-Terminal region of human p300.. Alternate names: P300, E1A-associated protein p300, P300 transcription coactivator homologueApplicationWB, IHC, ELISA. Species reactivity: Human
Catalog No. NC0759605
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.