Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA 531.1 Carbamate Mix, 100 μg/mL Each Component in Methanol, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017191 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7191 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7191 Supplier MilliporeSigma Supelco Supplier No. 46856U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EPA 531.1 Carbamate Mix may be utilized for identifying and quantifying carbamates in drinking water. The pre-requisite of EPA 531.1 method is the use of gas chromatography with a selective detector, gas chromatography/mass spectrometry or high performance liquid chromatography.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Concentration 100 μg/mL each component in methanol
Type Organic Pollutant Standard
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Aldicarb; Aldicarb-sulfone; Aldicarb-sulfoxide; Carbofuran; Carbofuran-3-hydroxy; 1-Naphthyl-N-methylcarbamate; Methiocarb; Methomyl; Oxamyl; Propoxur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.