Learn More
EPA 610 Polynuclear Aromatic Hydrocarbons Mix, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111017144 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
11-101-7144 1 mL
1 options
Catalog No. 11-101-7144 Supplier MilliporeSigma Supelco Supplier No. 4S8743
In Methanol: Methylene chloride (1:1) (Varied Conc.)
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Product Line TraceCERT
Type Food and Cosmetic Component Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Acenaphthene 1000 μg/mL; Acenaphthylene 2000 μg/mL; Anthracene 100 μg/mL; Benz[a]anthracene 100 μg/mL; Benzo[b]fluoranthene 200 μg/mL; Benzo[k]fluoranthene 100 μg/mL; Benzo[ghi]perylene 200 μg/mL; Benzo[a]pyrene 100 μg/mL; Chrysene 100 μg/mL; Dibenz[a,h]anthracene 200 μg/mL; Fluoranthene 200 μg/mL; Fluorene 200 μg/mL;Indeno[1,2,3-cd]pyrene 100 μg/mL; Naphthalene 1000 μg/mL; Phenanthrene 100 μg/mL; Pyrene 100 μg/mL
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC