Learn More
EPA 8240B/8260A Matrix Spike Mix, Certified Reference Material, 2,500 μg/mL Each Component in Methanol, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 2500 μg/mL each component in methanol |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Benzene; Chlorobenzene; 1,1-Dichloroethylene; Toluene; Trichloroethylene |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.