Learn More
EPA 8270 Ether/Phthalate Mix, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL each component in dichloromethane |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Benzyl butyl phthalate; Bis(2-chloroethoxy)methane; Bis(2-chloroethyl) ether; Bis(2-ethylhexyl) phthalate; 4-Bromodiphenyl ether; 4-Chlorodiphenyl ether; Bis-(2-chloroisopropyl) ether; Dibutyl phthalate; Diethyl phthalate; Dimethyl phthalate; Di-n-octyl phthalate |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.