Learn More
EPA HC Purgeable C, 2,000 μg/mL Each Component in Methanol, Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Ampule |
| Concentration | 2,000 μg/mL each component in methanol |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Bromomethane; Chloroethane; Chloromethane; Vinyl chloride |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.