Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPB41L2/4.1G Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560358
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0358 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0358 Supplier Bethyl Laboratories, Inc Supplier No. A301425AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The 4. 1 gene family encodes a group of multifunctional cytoskeletal proteins (4. 1R, 4. 1G, 4. 1N and 4. 1B), which are predomitly expressed in the nervous system. 4. 1G is a protein that stabilizes spectrin-actin interactions and is associated with hereditary elliptocytosis. Red blood cell 4. 1, designated 4. 1R, is a multifunctional protein that is essential for maintaining erythrocyte shape and membrane mechanical properties. Both 4. 1R and 4. 1G are distributed in a unique pattern in the cerebellum and are believed to modulate the membrane mechanical properties of neuronal cells by promoting fodrin/actin association. 4. 1N and 4. 1B, designated EPB41L1 and EPB41L3, respectively, are strongly expressed in the brain. Antibodies to 4. 1N have been reported to detect multiple forms, each enriched in postsynaptic density preparations relative to brain homogenate. Antibodies to 4. 1B have been reported to detect two forms.

Specifications

Antigen EPB41L2/4.1G
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene EPB41L2
Gene Accession No. O43491
Gene Alias EPB41L2; 4.1-G; erythrocyte membrane protein band 4.1-like 2; RP3-324N14.1; DKFZp781D1972; DKFZp781H1755; OTTHUMP00000040262; erythrocyte membrane protein band 4.1 like-protein 2
Gene Symbols EPB41L2
Host Species Rabbit
Immunogen Between 575 and 625
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2037
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.