Learn More
Invitrogen™ EPB41L5 Polyclonal Antibody

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human HEK293 whole cell. IHC: human liver cancer tissue, human Ovarian cancer tissue. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | EPB41L5 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | EPB41L5 |
| Gene Accession No. | Q9HCM4 |
| Gene Alias | Band 4.1-like protein 5; BE37; EPB41L5; erythrocyte membrane protein band 4.1 like 5; KIAA1548; YMO1; YRT; yurt homolog |
| Gene Symbols | EPB41L5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.