Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCAM Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7209294 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-558-555 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-558-555 Supplier Proteintech Group Inc Supplier No. 210501AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Epithelial cell adhesion molecule (EpCAM) is a transmembrane glycoprotein mediating Ca2+-independent homotypic cell-cell adhesion in epithelia. This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy.

Specifications

Antigen EPCAM
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EPCAM
Gene Accession No. P16422, Q99JW5
Gene Alias CD326, CO 17A, CO17 1A, EGP, EGP 2, EGP314, EGP34, EGP40, Ep CAM, EPCAM, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733 2, hEGP 2, hEGP314, KS 1/4 antigen, KS1/4, KSA, M1S2, M4S1, MIC18, MK 1, TACST 1, TACSTD1, TROP1
Gene Symbols EPCAM
Host Species Rabbit
Immunogen EPCAM Fusion Protein Ag15393
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17075, 4072
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.