Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2, Mouse anti-Human, Clone: F2-27, Millipore Sigma™

Catalog No. p-7162965 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS202625 25 μg
MABS2026100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS202625 Supplier MilliporeSigma Supplier No. MABS202625UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-EphA2, clone F2-27, Cat. No. MABS2026, is a mouse monoclonal antibody that detects Ephrin type-A receptor 2 and has been tested for use in Flow Cytometry, Immunoprecipitation, and Inhibition Assay.

Ephrin type-A receptor 2 (UniProt: P29317; also known as EC: 2.7.10.1, Epithelial cell kinase, Tyrosine-protein kinase receptor ECK) is encoded by the EPHA2 (also known as ECK) gene (Gene ID: 1969) in human. Ephrin type-A receptor 2 is a homodimeric single-pass type I membrane protein that is expressed in brain and glioma tissue and in glioma cell lines. It is also highly expressed in tissues that contain a high proportion of epithelial cells. Its levels are shown to be up-regulated by UV irradiation via a TP53-independent, MAPK-dependent mechanism. Ephrin type-A receptor 2 serves as a receptor tyrosine kinase that binds promiscuously membrane-bound ephrin-A family ligands residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. It is involved in the regulation of migration, integrin-mediated adhesion, and in proliferation and differentiation of cells. Ephrin type-A receptor 2 is phosphorylated on tyrosine residues upon binding and activation by EFNA1. Phosphorylation on Tyr 588 and Tyr 594 is reported to be essential for binding VAV2 and VAV3 while phosphorylation on Tyr 735 and Tyr 930 are required for binding PI3-kinase p85 subunit. It is synthesized with a signal peptide (aa 1-23) that is cleaved off in the mature form. The mature form has an extracellular domain (aa 24-537), a short transmembrane domain (aa 538-558), and a cytoplasmic domain (aa 559-976). Two isoforms of Ephrin type-A receptor 2 have been described that are produced by alternative splicing. EphA2 is reported to be overexpressed in skin tumors murine models and its overexpression may represent a compensatory feedback mechanism during tumorigenesis. (Ref.: Guo, H., et al. (2006). Cancer Res. 66(14); 7050-7058).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA2
Applications Flow Cytometry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone F2-27
Conjugate Unconjugated
Dilution Inhibition Analysis: A representative lot inhibited cancer cell invasion in a three-dimensional spheroid formation assay. (Tanaka, T., et. al. (2017). Transl Oncol. 10(4):476-484).Immunoprecipitation Analysis: A representative lot detected EphA2 in Immunoprecipitation applications (Tanaka, T., et. al. (2017). Transl Oncol. 10(4):476-484).Flow Cytometry Analysis: A representative lot detected EphA2 in Flow Cytometry applications (Tanaka, T., et. al. (2017). Transl Oncol. 10(4):476-484).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ephrin type-A receptor 2;EC: 2.7.10.1;Epithelial cell kinase;Tyrosine-protein kinase receptor ECK
Gene Symbols EPHA2;ECK
Host Species Mouse
Immunogen KP-2, KP-3, SUIT-2, and MIAPaCa-2 cells.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_004422
Test Specificity Clone F2-27 specifically detects Ephrin type -A receptor 2 in human.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.