Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHA2 Mouse anti-Human, Unconjugated, Clone: 1A9C3, Proteintech
SDP

Catalog No. 502239016 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9016 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9016 Supplier Proteintech Group Inc Supplier No. 667361IG150UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The EPHA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHA2. This antibody recognizes human antigen. The EPHA2 antibody has been validated for the following applications: FC, WB, ELISA analysis.

Specifications

Antigen EPHA2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 1A9C3
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 1969
Target Species Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.