Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547017
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547017 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547017 Supplier Invitrogen™ Supplier No. PA547017
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 5% cross-reactivity with recombinant rat (rr) EphB1, recombinant mouse (rm) EphA8, rmEphA6, rmEphB6, rmEphA3, rmEphA4, rmEphA7, rrEphA5, recombinant human EphA1, rmEphA2 and rmEphB3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

EPHB2 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphB2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and no preservative
Gene EPHB2
Gene Accession No. P29323, P54763
Gene Alias CAPB; cEK5; chicken embryo kinase 5; chicken embryo kinase 5 protein; developmentally-regulated Eph-related tyrosine kinase; DRT; EFL6; EFNB3; EK5; ELK; ELK-related tyrosine kinase; embryo kinase 5 protein CEK5; EPH B2; EPH receptor B2; eph tyrosine kinase 3; EPHB2; EphB2/CTF1; EphB2/CTF2; EPHB3; EPH-like kinase 5; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 2; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPHT3; EPLG8; EPTH3; ERK; ETECK; ETK2; HEK2; Hek5; HEK-6; LERK8; MGC87492; NET; Neural kinase; Nuk; Nuk receptor tyrosine kinase; PCBC; Prkm5; protein-tyrosine kinase HEK5; Qek5; Renal carcinoma antigen NY-REN-47; RGD1564232; Sek3; TYRO5; TYRO6; Tyrosine-protein kinase receptor EPH-3; tyrosine-protein kinase receptor SEK-3; Tyrosine-protein kinase TYRO5
Gene Symbols EPHB2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Val27-Lys548.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13844, 2048
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.