Learn More
Invitrogen™ Ephrin A1 Recombinant Rabbit Monoclonal Antibody (067), Invitrogen™
Shop All Thermo Scientific ProductsDescription
Specificity: Rat Ephrin-A1/EFNA1
Specifications
Specifications
| Antigen | Ephrin A1 |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 067 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | EFNA1 |
| Gene Accession No. | P97553 |
| Gene Alias | AI325262; B61; ECKLG; EFL1; Efna1; Epgl1; EPH-related receptor tyrosine kinase ligand 1; ephrin A1; ephrin-A1; Ephrin-A1, secreted form; Epl1; EPLG1; Immediate early response protein B61; Lerk1; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; TNFAIP4; tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.