Learn More
Invitrogen™ Ephrin A3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total Ephrin A3 / EFNA3 protein.
Specifications
Specifications
| Antigen | Ephrin A3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | EFNA3 |
| Gene Accession No. | O08545, P52797 |
| Gene Alias | AW494418; EFL2; EFL-2; EFNA3; EHK1 ligand; EHK1-L; EPH-related receptor tyrosine kinase ligand 3; ephrin A3; ephrin-A3; Epl3; EPLG3; Lerk3; LERK-3; ligand of eph-related kinase 3 |
| Gene Symbols | EFNA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.