Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ephrin B1 Monoclonal Antibody (25H11), eBioscience™

Catalog No. 14530080
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-530-080 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-530-080 Supplier Invitrogen™ Supplier No. 14530080
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody 25H11 recognizes mouse Ephrin B1, a 47 kDa membrane protein. Ephrin B1 belongs to a family of ephrins composed of type A (linked to the cell membrane through a glycosylphosphatidyl inositol group) or type B (transmembrane) proteins. The type B ephrins consist of 3 members (B1, B2, and B3) each of which contains a cytoplasmic domain containing tyrosine residues that can be phosphorylated. Ephrin B and ephrin B receptors (Eph B) can interact to potentiate bidirectional downstream signaling between cells expressing either or both the ligand and receptor. Ephrins have been shown to play a role in neural crest cell development through both neurogenesis and neural migration, as well as development of kidney and heart cyto-architecture. Overexpression of type B ephrins in cancer cells is associated with increased vascularity and highly invasive tumors. Applications Reported: This 25H11 antibody has been reported for use in immunohistochemical staining of frozen tissue sections and immunoprecipitation. Applications Tested: This 25H11 antibody has been tested immunohistochemistry on frozen mouse kidney tissue sections at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. Binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. The signaling pathway downstream of the receptor is referred to as forward signaling while the signaling pathway downstream of the ephrin ligand is referred to as reverse signaling. Binds to receptor tyrosine kinase including EPHA4, EPHA3 and EPHB4. Together with EPHB4 plays a central role in heart morphogenesis and angiogenesis through regulation of cell adhesion and cell migration. EPHB4-mediated forward signaling controls cellular repulsion and segregation from EFNB2-expressing cells. May play a role in constraining the orientation of longitudinally projecting axons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin B1
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 25H11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene EFNB1
Gene Accession No. P52795
Gene Alias CEK5 ligand; CEK5 receptor ligand; CEK5-L; CEL5-L; CFND; CFNS; EFB1; EFL3; EFL-3; EFL6; EFNB1; EFNB3; ELK ligand; ELK-L; EPH-related receptor tyrosine kinase ligand 2; ephrin B1; Ephrin B3; EphrinB1; ephrin-B1; Ephrin-B1 C-terminal fragment; Ephrin-B1 CTF; Ephrin-B1 ICD; Ephrin-B1 intracellular domain; Ephrin-B1 precursor; Epl2; EPLG2; EPLG8; kinase ephrin-B1; Lerk2; LERK-2; LERK8; Stimulated by retinoic acid gene 1 protein; Stra1; STRA1 protein; transmembrane ligand
Gene Symbols EFNB1
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13641
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.