Learn More
Invitrogen™ Ephrin B2 Polyclonal Antibody
Description
Recommended positive controls: HeLa, mouse brain, rat brain. Predicted reactivity: Mouse (96%), Rat (97%), Dog (95%), Cat (95%), Pig (95%), Chicken (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Ephrin B2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Efnb2 |
| Gene Accession No. | P52799, P52800 |
| Gene Alias | EFNB2; efnb2a; Elf2; ELF-2; EPH-related receptor tyrosine kinase ligand 5; ephrin B2; ephrin B2a; ephrin-B2; Ephrin-B2a; Epl5; EPLG5; HTK ligand; Htkl; HTK-L; id:ibd5072; Lerk5; LERK-5; ligand of eph-related kinase 5; MGC126226; MGC126227; MGC126228; NLERK-1 |
| Gene Symbols | Efnb2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.