Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPI64 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501568118
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8118 Supplier Bethyl Laboratories, Inc Supplier No. A304587A
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The function of this protein remains unknown.

Specifications

Antigen EPI64
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TBC1D10A
Gene Accession No. P58802, Q9BXI6
Gene Alias AC004997.C22.2, dJ130H16.1, dJ130H16.2, EBP50-PDX interactor of 64 kDa, EBP50-PDZ interactor of 64 kD, EPI64, EPI64 protein, rab27A-GAP-alpha, TBC1 domain family, member 10, TBC1 domain family, member 10A, TBC1D10
Gene Symbols TBC1D10A
Host Species Rabbit
Immunogen Between 458 and 508
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103724, 83874
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.